Skip to Content
Merck
All Photos(1)

Key Documents

914312

Sigma-Aldrich

N3-TFBA-O2Oc

≥98%

Synonym(s):

2-(2-(2-(4-Azido-2,3,5,6-tetrafluorobenzamido)ethoxy)ethoxy)acetic acid, 4-Azido-2,3,5,6-tetrafluorobenzoyl-AEEA, 8-(4-Azido-2,3,5,6-tetrafluorobenzoyl-amino)-3,6-dioxaoctanoic acid, ATFB-AEEA, ATFB-Ado, ATFB-Adoa

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12F4N4O5
CAS Number:
Molecular Weight:
380.25
UNSPSC Code:
12352200

Assay

≥98%

form

powder

availability

available only in USA

mp

88-89 °C

storage temp.

2-8°C

Application

N3-TFBA-O2Oc is a photoactivatable crosslinker with a carboxylic acid on one end and a perfluorophenyl azide on the other. Perfluorophenyl azides react upon irradiation (258-280 nm), providing a means of spatial and temporal control when incorporated into chemical tools for biological applications, surface modifications, or nanomaterials synthesis.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Self-react. C - Skin Irrit. 2

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service