914029
IEICO-4F
≥99%
Synonym(s):
2,2′-((2Z,2′Z)-(((4,4,9,9-tetrakis(4-hexylphenyl)-4,9-dihydro-sindaceno[1,2-b:5,6-b′]dithiophene-2,7-diyl)bis(4-((2-ethylhexyl)oxy)thiophene-5,2-diyl))bis(methanylylidene))bis(5,6-difluoro-3-oxo-2,3-dihydro-1H-indene-2,1-diylidene))dimalononitrile
Select a Size
Select a Size
About This Item
Recommended Products
description
Band gap: Eg=1.29 eV
Quality Level
Assay
≥99%
form
solid
color
dark
Orbital energy
HOMO -5.49 eV
LUMO -4.19 eV
SMILES string
Fc1cc2c(cc1F)C(=C(C#N)C#N)\C(=C/c3[s]c(c(c3)OCC(CCCC)CC)c4[s]c5c(c4)C(c8c5cc9c(c8)c%10[s]c(cc%10C9(c%15ccc(cc%15)CCCCCC)c%14ccc(cc%14)CCCCCC)c%11[s]c(cc%11OCC(CCCC)CC)\C=C%12/C(=O)c%13c(cc(c(c%13)F)F)C/%12=C(C#N)C#N)(c7ccc(cc7)CCCCCC)c6ccc(cc6)CCCCCC)\C2=
InChI
1S/C114H114F4N4O4S4/c1-9-17-23-27-33-73-37-45-79(46-38-73)113(80-47-39-74(40-48-80)34-28-24-18-10-2)93-57-90-94(58-89(93)109-95(113)63-103(129-109)111-101(125-69-71(15-7)31-21-13-5)55-83(127-111)53-91-105(77(65-119)66-120)85-59-97(115)99(117)61-87(85)107(
InChI key
ZDIRJORRKGPNQG-ZTVJXLAYSA-N
Related Categories
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service