913944
Silica dioxide-electrospun milled nanofibers
Synonym(s):
Nanofibrous inorganic powder structure, SiO2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
nanofiber
Quality Level
fiber diam. × L
100-300 nm × 2-10 μm , Structure: random fiber mesh
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Related Categories
Application
Silica dioxide electrospun milled nanofibers can absorb significantly more water than commercially aailable silicagel of the same mesoporous character. Applications include adsorbent of water and other polar sorbents, catalyst carrier, filtration, separation, li-ion battery separators, and sensors.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Review discusses recent progress and challenges in ORR catalyst development for PEMFC applications.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service