Skip to Content
Merck
All Photos(1)

Key Documents

913685

Sigma-Aldrich

Zirconium oxide milled nanofibers

Synonym(s):

Nanofibrous inorganic powder structure, Zirconium Dioxide Nanofibers ZrO2

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€162.00
10 G
€281.00

€162.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€162.00
10 G
€281.00

About This Item

Linear Formula:
ZrO2
CAS Number:
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

€162.00


Please contact Customer Service for Availability

Request a Bulk Order

form

nanofiber

Quality Level

fiber diam. × L

200-800 nm ± 100 nm × 2-10 μm , Structure: Polycystalline nanofibers

surface area

20-30 m2/g

primary crystallite size

(monoclinic and tetragonal
Typical Size of Crystallites: ∼8-13 nm)

SMILES string

[Zr](=O)=O

InChI key

MCMNRKCIXSYSNV-UHFFFAOYSA-N

Application

Zirconium oxide has applications in electro ceramics, catalysts, catalyst support, piezoelectric crystals, oxygen sensors, abrasives, thermal barrier coating, fireproof materials, ion conductors, buffer layers for growing superconductors, materials for high temperature chemical processes, and inorganic separators.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Review discusses recent progress and challenges in ORR catalyst development for PEMFC applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service