Skip to Content
Merck
All Photos(1)

Key Documents

912948

Sigma-Aldrich

Ir[FCF3(CF3)ppy]2(dtbbpy)PF6

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€147.00
500 MG
€948.00

€147.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€147.00
500 MG
€948.00

About This Item

Empirical Formula (Hill Notation):
C44H34F20IrN4P
CAS Number:
Molecular Weight:
1221.93
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€147.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

mp

378 °C

SMILES string

[Ir+].FC(F)(F)c5cnc(cc5)c6c(cc(cc6)C(F)(F)F)F.FC(F)(F)c3cnc(cc3)c4c(cc(cc4)C(F)(F)F)F.F[P-](F)(F)(F)(F)F.n1c(cc(cc1)C(C)(C)C)c2nccc(c2)C(C)(C)C

InChI key

HNVAXJZTSVKZHB-UHFFFAOYSA-N

Application

Ir[FCF3(CF3)ppy]2(dtbbpy)PF6 is a versatile photocatalyst that can be used for a variety of synthetic transformations including the α-arylation of alcohols.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jack Twilton et al.
Angewandte Chemie (International ed. in English), 57(19), 5369-5373 (2018-03-01)
The combination of nickel metallaphotoredox catalysis, hydrogen atom transfer catalysis, and a Lewis acid activation mode, has led to the development of an arylation method for the selective functionalization of alcohol α-hydroxy C-H bonds. This approach employs zinc-mediated alcohol deprotonation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service