Skip to Content
Merck
All Photos(1)

Key Documents

912697

Sigma-Aldrich

Poly(acrylamide) solution

average Mw ~10,000, 50 wt. % in H2O

Synonym(s):

PAM

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€164.00

€164.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€164.00

About This Item

Linear Formula:
(C3H5NO)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€164.00


Please contact Customer Service for Availability

Request a Bulk Order

form

viscous liquid

Quality Level

mol wt

average Mw ~10,000

concentration

50 wt. % in H2O

color

colorless to light brown

SMILES string

NC(=O)C=C

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)

InChI key

HRPVXLWXLXDGHG-UHFFFAOYSA-N

Application

Poly(acrylamide) is a water-soluble polymer made up of acrylamide subunits. Polyacrylamides have been reported to be protein resistant and biocompatible. Polyacrylamides have been used as carriers for delivery of drugs and bioactive compounds, or to as tissue engineering materials. Additionally, polycacrylamides have been used to increase the viscosity of water and facilitate the flocculation of particles present in water.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent Applications of Polyacrylamide as Biomaterials.
Yang T H, et al.
Materials Science, 100(1) (2008)
James E Woodrow et al.
Journal of agricultural and food chemistry, 56(8), 2773-2779 (2008-03-21)
Linear anionic polyacrylamide (PAM) has been used in irrigation practices as a flocculating agent to minimize water losses through seepage in earthen canals. The stability of PAM is of concern because of the possibility of acrylamide (AMD) monomer release during

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service