Skip to Content
Merck
All Photos(1)

Key Documents

912603

Sigma-Aldrich

Di-n-propylammonium bromide

Synonym(s):

1-Propanamine, N-propyl-, DNPRAB, Greatcell Solar®, Hydrobromide (8CI) dipropylammonium bromide, Hydrobromide (9CI) dipropylamine

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€81.60
50 G
€305.00

€81.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€81.60
50 G
€305.00

About This Item

Empirical Formula (Hill Notation):
C6H16BrN
CAS Number:
Molecular Weight:
182.10
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

€81.60


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

Quality Level

color

white

SMILES string

[Br-].[N+H2](CCC)CCC

InChI

1S/C6H15N.BrH/c1-3-5-7-6-4-2;/h7H,3-6H2,1-2H3;1H

InChI key

FZJRCNLPISUZCV-UHFFFAOYSA-N

Application

Organohalide based perovskites have emerged as an important class of material for solar cell applications.[1][2][3] The variations/substitution in organohalide cations and anions is employed for the optimization of the band gap, carrier diffusion length, and power conversion efficiency of perovskites based solar cells.

Legal Information

Product of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High Efficiency and High Open Circuit Voltage in Quasi 2D Perovskite Based Solar Cells.
Cohen B E, et al.
Advances in Functional Materials, 27(5), 1604733-1604733 (2017)
Lingling Mao et al.
Journal of the American Chemical Society, 139(14), 5210-5215 (2017-03-18)
Hybrid inorganic-organic perovskites are developing rapidly as high performance semiconductors. Recently, two-dimensional (2D) perovskites were found to have white-light, broadband emission in the visible range that was attributed mainly to the role of self-trapped excitons (STEs). Here, we describe three
Eran Edri et al.
The journal of physical chemistry letters, 4(6), 897-902 (2013-03-21)
Mesoscopic solar cells, based on solution-processed organic-inorganic perovskite absorbers, are a promising avenue for converting solar to electrical energy. We used solution-processed organic-inorganic lead halide perovskite absorbers, in conjunction with organic hole conductors, to form high voltage solar cells. There

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service