Skip to Content
Merck
All Photos(1)

Key Documents

910414

Sigma-Aldrich

1,10-Decanediol dimethacrylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€193.00

€193.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€193.00

About This Item

Empirical Formula (Hill Notation):
C18H30O4
CAS Number:
Molecular Weight:
310.43
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€193.00


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

contains

<300 ppm HQ as stabilizer

color

colorless

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCCCCCCCCCCOC(=O)C(C)=C

InChI

1S/C18H30O4/c1-15(2)17(19)21-13-11-9-7-5-6-8-10-12-14-22-18(20)16(3)4/h1,3,5-14H2,2,4H3

InChI key

LRZPQLZONWIQOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,10-Decanediol dimethacrylate is a crosslinker with hydrophobic spacer.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

>230.9 °F

Flash Point(C)

> 110.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service