Skip to Content
Merck
All Photos(2)

Key Documents

909734

Sigma-Aldrich

1-Phenyl-2-((trifluoromethyl)sulfonyl)propan-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9F3O3S
CAS Number:
Molecular Weight:
266.24
UNSPSC Code:
12352101

form

crystals

reaction suitability

reaction type: C-C Bond Formation

functional group

fluoro
ketone
phenyl
sulfone

storage temp.

−20°C

Application

1-Phenyl-2-((trifluoromethyl)sulfonyl)propan-1-one is part of a suite of reagents developed by the Li group for late-stage functionalization of pharmaceutically relevant compounds. The reagents serve as precursors to alkyl radicals generated under light mediated, redox-neutral and catalyst free conditions for use in the Minisci alkylation of aromatics.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Liu et al.
Journal of the American Chemical Society, 139(40), 14315-14321 (2017-10-03)
The Minisci alkylation is useful to functionalize aromatics via alkyl radical addition. Current approaches to prepare alkyl radicals follow either oxidative or reductive pathways from various functional groups. Developing new strategy beyond these traditional methods remains elusive yet highly significant.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service