907863
(S)-2-((3-(tert-butyl)-2-hydroxybenzyl)amino)-N,N,3-trimethylbutanamide
≥95%
Synonym(s):
Hoveyda aminophenol catalyst
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H30N2O2
CAS Number:
Molecular Weight:
306.44
UNSPSC Code:
12352111
Assay
≥95%
form
powder or chunks
mp
97-99 °C
storage temp.
2-8°C
Application
(S)-2-((3-(tert-butyl)-2-hydroxybenzyl)amino)-N,N,3-trimethylbutanamide is an aminophenol organocatalyst developed by the Hoveyda lab for enantioselective addition of allylboron compounds to imines, ketones, and other carbonyls, including polyfluorinated substrates.
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Farid W van der Mei et al.
Angewandte Chemie (International ed. in English), 55(15), 4701-4706 (2016-03-11)
Catalytic allylboron additions to aldimines are presented for which small amounts of Zn(OMe)2 serve as the co-catalyst to accelerate allyl exchange and 1,3-borotropic shift processes. Low-yielding and moderately α- and diastereoselective reactions are thus turned into highly efficient γ-, diastereo-
Hao Wu et al.
Journal of the American Chemical Society, 136(10), 3780-3783 (2014-03-05)
A practical catalytic method for enantioselective addition of an allene unit to aldimines is disclosed. Transformations are promoted by an in-situ-generated B-based catalyst that is derived from a simple, robust, and readily accessible (in multigram quantities) chiral aminoalcohol. A range
Daniel W Robbins et al.
Angewandte Chemie (International ed. in English), 55(33), 9610-9614 (2016-06-09)
A set of broadly applicable methods for efficient catalytic additions of easy-to-handle allyl-B(pin) (pin=pinacolato) compounds to ketones and acyclic α-ketoesters was developed. Accordingly, a large array of tertiary alcohols can be obtained in 60 to >98 % yield and up to
KyungA Lee et al.
Nature chemistry, 8(8), 768-777 (2016-07-22)
Organofluorine compounds are central to modern chemistry, and broadly applicable transformations that generate them efficiently and enantioselectively are in much demand. Here we introduce efficient catalytic methods for the addition of allyl and allenyl organoboron reagents to fluorine-substituted ketones. These
Daniel L Silverio et al.
Nature, 494(7436), 216-221 (2013-02-15)
The discovery of catalysts that can be used to synthesize complex organic compounds by enantioselective transformations is central to advances in the life sciences; for this reason, many chemists aim to discover catalysts that allow for preparation of chiral molecules
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service