Skip to Content
Merck
All Photos(3)

Key Documents

904104

Sigma-Aldrich

CoCl2(PCy3)2

Synonym(s):

Dichlorobis(tricyclohexylphosphine)cobalt(II)

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€62.20

€62.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€62.20

About This Item

Empirical Formula (Hill Notation):
C36H66Cl2CoP2
CAS Number:
Molecular Weight:
690.70
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€62.20


Please contact Customer Service for Availability

Request a Bulk Order

form

powder or crystals

reaction suitability

core: cobalt
reagent type: catalyst

mp

>300 °C

SMILES string

[Co](Cl)Cl.P(C6CCCCC6)(C5CCCCC5)C4CCCCC4.P(C3CCCCC3)(C2CCCCC2)C1CCCCC1

InChI

1S/2C18H33P.2ClH.Co/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*16-18H,1-15H2;2*1H;/q;;;;+2/p-2

InChI key

XTBDMUPZCVAWRK-UHFFFAOYSA-L

Application

This cobalt catalyst can be used in conjunction with the photocatalyst (Ir[dF(CF3)ppy]2(dtbpy))PF6(747793) for the [2+2+2] alkyne cycloaddition.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyle E Ruhl et al.
Journal of the American Chemical Society, 138(48), 15527-15530 (2016-12-10)
The ability to exert spatial and temporal control over a transition-metal catalyst offers diverse opportunities for the fabrication of functional materials. Using an external stimulus such as visible light to toggle a catalyst between an active and dormant state has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service