Skip to Content
Merck
All Photos(2)

Key Documents

901725

Sigma-Aldrich

1-(Tert-butyl) 2-methyl (2S,4R)-4-((4-bromo-7-fluoroisoindoline-2-carbonyl)oxy)pyrrolidine-1,2-dicarboxylate

Synonym(s):

Aryl halide chemistry informer library compound X4

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€167.00

€167.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€167.00

About This Item

Empirical Formula (Hill Notation):
C20H24BrFN2O6
CAS Number:
Molecular Weight:
487.32
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€167.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder or crystals

mp

157.61 °C

Application

This product is Informer compound X4 of the Aryl halide chemistry informer library developed by chemists at Merck & Co. Inc., Kenilworth, NJ, U.S., which contains 18 drug-like molecules representative of those encountered in complex synthesis. By screening a new reaction against the informer library, chemists can directly compare and analyse a reaction′s successes and shortcomings among different methods and various research teams. It may also be used to facilitate deeper method development for performance or utility.

Caution

Not fully tested

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas J Greshock et al.
Angewandte Chemie (International ed. in English), 55(44), 13714-13718 (2016-10-22)
The reactivity of a representative set of 17 organozinc pivalates with 18 polyfunctional druglike electrophiles (informers) in Negishi cross-coupling reactions was evaluated by high-throughput experimentation protocols. The high-fidelity scaleup of successful reactions in parallel enabled the isolation of sufficient material
Peter S Kutchukian et al.
Chemical science, 7(4), 2604-2613 (2016-04-21)
Major new advances in synthetic chemistry methods are typically reported using simple, non-standardized reaction substrates, and reaction failures are rarely documented. This makes the evaluation and choice of a synthetic method difficult. We report a standardized complex molecule diagnostic approach

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service