Skip to Content
Merck
All Photos(1)

Key Documents

901441

Sigma-Aldrich

Tri-tert-butylphosphine solution

1.0 M in 2-methyltetrahydrofuran

Synonym(s):

P(t-Bu)3

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€258.00

€258.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€258.00

About This Item

Empirical Formula (Hill Notation):
C12H27P
CAS Number:
Molecular Weight:
202.32
MDL number:
UNSPSC Code:
12352128
NACRES:
NA.22

€258.00


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

Quality Level

reaction suitability

reagent type: ligand
reaction type: Addition Reactions

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Negishi Coupling

reagent type: ligand
reaction type: Sonogashira Coupling

reagent type: ligand
reaction type: Stille Coupling

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

concentration

1.0 M in 2-methyltetrahydrofuran

density

0.853 g/mL

functional group

phosphine

SMILES string

CC(C)(C)P(C(C)(C)C)C(C)(C)C

InChI

1S/C12H27P/c1-10(2,3)13(11(4,5)6)12(7,8)9/h1-9H3

InChI key

BWHDROKFUHTORW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Valuable ligand for several coupling reactions. This product is offered as a solution in 2-methyltetrahydrofuran for more convenient handling.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

6.8 °F

Flash Point(C)

-14 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service