Skip to Content
Merck
All Photos(1)

Documents

901268

Sigma-Aldrich

Pure Strip

stabilized sulfuric acid-hydrogen peroxide compound

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

form

liquid

General description

Pure Strip is suitable for stripping both positive and negative photoresists in addition to other organic compounds in a variety of semiconductor, photomask, and IC photolithography compounds. High yields can be achieved due to the high purity/low particulate composition of Pure Strip.
Advantages Pure Strip include negligible attack on exposed metal surfaces, including aluminum, vs. other acidic formulations, residue-free rinsing, and extended bath life (minimum of five days at room temperature). Pure Strip is ready to use and requires no mixing.

Application

Pure Strip may be used at room temperature or at elevated temperature. Higher temperatures will increase the activity but decrease the bath life (1 day at 60-80 °C). Substrates are stripped of photoresist and cleaned effectively with minimal attack on aluminum (approximately 35 Angstroms/minute at room temperature) and negligible attack on other metals and alloys such as titanium, Ti-tungsten, copper, tantalum silicide, and ITO.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service