Skip to Content
Merck
All Photos(1)

Documents

901257

Sigma-Aldrich

2-Hydrazinoquinoline

≥98%

Synonym(s):

2-Hydrazineylquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9N3
CAS Number:
Molecular Weight:
159.19
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98%

form

powder

mp

140-145 °C

InChI

1S/C9H9N3/c10-12-9-6-5-7-3-1-2-4-8(7)11-9/h1-6H,10H2,(H,11,12)

InChI key

QMVCLSHKMIGEFN-UHFFFAOYSA-N

Application

2-Hydrazinoquinoline is a hydrazine reagent mainly used in the preparation of hydrazone derivatives of carbonyl compounds. This product was previously listed as CDS000062.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The magnetic exchange interaction via N?H⋅⋅⋅ O-bonding in copper (II) complex with 1-phenyl-3-methyl-4-formylpyrazol-5-one 2-quinolylhydrazone.
Levchenkov SI et al.
Inorgorganica Chimica Acta, 405, 169-175 (2013)
Jung H Suh et al.
Chemistry and physics of lipids, 194, 101-109 (2015-09-27)
A new technique for quantifying sphingosine phosphate lyase activity in biological samples is described. In this procedure, 2-hydrazinoquinoline is used to convert (2E)-hexadecenal into the corresponding hydrazone derivative to improve ionization efficiency and selectivity of detection. Combined utilization of liquid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service