Skip to Content
Merck
All Photos(1)

Key Documents

900898

Sigma-Aldrich

Hyflon® AD 40L S

Synonym(s):

Amorphous perfluoropolymer solution, P(TTD-TFE), Poly(tetrafluoroethylene-co-2,2,4-trifluoro-5-trifluoromethoxy-1,3-dioxole), TTD-TFE copolymers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4F6O3)n(C2F4)m
CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.23

description

Polymer content: Hyflon® AD 40L 9.0 ± 0.5 wt%
Solution in perfluorinated solvent (Galden® D02TS)

form

liquid

composition

TTD content, 40 ±5 mol % by

transmittance

>95% (in UV-Vis range)

viscosity

600 cP(25 °C, Brookfield)

General description

  • Hyflon® AD 40L S is a commerical grade Hyflon® AD 40H perfluoropolymer solution in a perfluorinated solvent. This can be considered ready-to-use solution suitable for application in the most demanding solution process technologies. Amorphous glassy polymer structure.
  • Solubility in perfluorinated solvents.
  • Low dielectric constant of 2.0 at 1 kHz.
  • Excellent optical properties:
a) High UV transmittance.
b) Low refractive index.
Hyflon® AD can provide uniform and thin films or coatings (down to submicron thickness if required) by casting or coating techniques and thermal treatments, well below the typical baking temperatures of semicrystalline perflouropolymers.

Key features of Hyflon® AD amorphous perfluoropolymers:
  • High temperature stability.
  • Good chemical resistance.
  • Excellent hydrophobicity.
  • Very low surface energy.
  • Low dielectric constant.
  • Very high gas permeability and selectivity.
  • Outstanding optical properties (e.g. high UV transmittance, low refractive index).

Examples of typical applications include:
  • Protective coatings.
  • Interlayer dielectric films and pellicles for electronics.

Legal Information

Galden is a registered trademark of Solvay Group
Hyflon is a registered trademark of Solvay Group

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service