Skip to Content
Merck
All Photos(2)

Key Documents

900809

Sigma-Aldrich

1-Butyl-1-methylpyrrolidinium tetrafluoroborate

98%, H2O <500 ppm

Synonym(s):

N-n-Butyl-N-methylpyrrolidinium tetrafluoroborate, Butylmethylpyrrolidinium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20BF4N
CAS Number:
Molecular Weight:
229.07
MDL number:
UNSPSC Code:
12352100

grade

battery grade

Assay

98%

form

powder

composition

H2O, <500 ppm

impurities

≤500 ppm H2O

mp

150 °C

application(s)

battery manufacturing

SMILES string

F[B-](F)(F)F.CCCC[N+]1(C)CCCC1

InChI

1S/C9H20N.BF4/c1-3-4-7-10(2)8-5-6-9-10;2-1(3,4)5/h3-9H2,1-2H3;/q+1;-1

InChI key

PGCVCJOPLBWQHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Butyl-1-methylpyrrolidinium tetrafluoroborate, a green solvent, is an ionic liquid that can be used in the formation of an electrolytic solution. An electrolytic solution can be used in a variety of applications such as double-layered capacitors, polymeric batteries, and solid-state actuators.[1][2][3]
Ionic liquids (ILs) are molten salts with melting points lower than 100 °C. They usually consist of pair of organic cation and anion. ILs exhibit unique properties such as nonvolatility, high thermal stability, and high ionic conductivity and find applications as electrolytes in lithium/sodium ion batteries and dye-sensitized solar cells. They are also used as media for synthesis of conducting polymers and intercalation electrode materials.[4][5][6]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantum chemical studies on the simultaneous interaction of thiophene and pyridine with ionic liquid
Anantharaj R and Banerjee T
AIChE Journal, 57(3), 749-764 (2011)
Influence of the salt concentration on the electrochemical performance of electrodes for polymeric batteries
Gerlach P, et al.
Electrochimica Acta, 306(3), 610-616 (2019)
The effect of ionic liquid type on the properties of hydrogenated nitrile elastomer/hydroxy-functionalized multi-walled carbon nanotube/ionic liquid composites
Likozar B
Soft Matter, 7(3), 970-977 (2011)
Masayoshi Watanabe et al.
Chemical reviews, 117(10), 7190-7239 (2017-01-14)
Ionic liquids (ILs) are liquids consisting entirely of ions and can be further defined as molten salts having melting points lower than 100 °C. One of the most important research areas for IL utilization is undoubtedly their energy application, especially
Ionic liquids and their solid-state analogues as materials for energy generation and storage.
MacFarlane DR, et al.
Nature Reviews. Materials, 1, 15005-15005 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service