Skip to Content
Merck
All Photos(2)

Documents

900663

Sigma-Aldrich

Methoxy poly(ethylene glycol)-b-poly(D,L-lactide)

10k-10k

Synonym(s):

mPEG-b-PLA, mPEG-PLA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO[CH(CH3)COO]m[CH2CH2O]nCH3
UNSPSC Code:
12162002
NACRES:
NA.23

form

crystals

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Biocompatible, amphiphilic block copolymer composed of a hydrophilic PEG block and a hydrophobic poly(D,L-lactide) (PLA) block. These materials have been used in control release and nanoparticle formulation for drug encapsulation and delivery applications. Well-defined materials with varying properties can be prepared by controlling the relative length of each polymer block. Hydroxyl termination allows for facile further chemical modification of these materials.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yiguang Wang et al.
Pharmaceutical research, 27(9), 1861-1868 (2010-06-19)
To develop an efficient tumor vasculature-targeted polymeric micelle delivery system for combretastatin A4 (CA4), a novel antivascular agent. CA4-loaded micelles were prepared from poly (ethylene glycol)-b-poly (d, l-lactide) copolymers. RGD peptides that target integrins alphavbeta3 and alphavbeta5, markers of angiogenic
Yuan Zhang et al.
Pharmaceutical research, 28(5), 1167-1178 (2011-02-23)
Somatostatin analogue octreotide (OCT)-modified PEG-b-PLA micelles were constructed to bind to somatostatin receptors (SSTRs) overexpressed on tumor cells for enhanced intracellular drug delivery and improved therapeutic efficacy for malignant tumors. Copolymers conjugated with octreotide (OCT-PEG₆₀₀₀-b-PLA₅₀₀₀) were synthesized. The fluorescent probe
Zahra Daman et al.
Pharmaceutical research, 32(11), 3756-3767 (2015-08-01)
Resistance to gemcitabine in pancreatic cancer (PC) may account for the failure of conventional treatments. Recently, salinomycin (SAL) has been identified as selective inhibitor of cancer stem cells (CSCs). In our study, we aimed to deliver SAL to gemcitabine-resistant PC
Ho-Chul Shin et al.
Journal of controlled release : official journal of the Controlled Release Society, 140(3), 294-300 (2009-05-05)
Current clinical and preclinical anticancer formulations are limited by their use of toxic excipients and stability issues upon combining different drug formulations. We have found that poly(ethylene glycol)-block-poly(d,l lactic acid) (PEG-b-PLA) micelles can deliver multiple poorly water-soluble drugs at clinically

Articles

The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service