Skip to Content
Merck
All Photos(1)

Key Documents

900617

Sigma-Aldrich

(4-(Hydroxymethyl)phenyl)(4-(prop-2-yn-1-yloxy)phenyl)methanone

≥95%

Synonym(s):

Hydroxyl benzophenone alkyne, Probe building block

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O3
CAS Number:
Molecular Weight:
266.29
UNSPSC Code:
12352200

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: click chemistry

storage temp.

−20°C

Application

(4-(Hydroxymethyl)phenyl)(4-(prop-2-yn-1-yloxy)phenyl)methanone is used for chemical probe synthesis, this trifunctional building block contains a light-activated benzophenone, alkyne tag, and hydroxyl synthetic handle. When appended to a ligand or pharmacophore through its hydroxyl linker, this building block allows for UV light-induced covalent modification of a biological target with potential for downstream applications via the alkyne tag. Use alone or in parallel with other multi-functional building blocks to discover the optimal probe for your chemical biology experiments.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In partnership with Pfizer chemists, we have compiled a collection of trifunctional building blocks to enable development of probes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service