Skip to Content
Merck
All Photos(1)

Key Documents

900486

Sigma-Aldrich

Gold nano-urchins

70 nm, maleimide functionalized, conjugation kit

Synonym(s):

Maleimide-activated gold nanourchins, Au nanoparticles, Gold nanoparticles, Urchins

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€372.00

€372.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€372.00

About This Item

UNSPSC Code:
12141717
NACRES:
NA.23

€372.00


Please contact Customer Service for Availability

Request a Bulk Order

form

nanourchin

shelf life

12 mo. at -20 °C (If stored unopened and as specified, are stable for at least 12 months)

storage condition

protect from light

particle size

70 nm

λmax

600 nm

functional group

maleimide

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of CytoDiagnostics, Inc.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Drs. Fernandes and Baptista discuss gold nanoparticles synthesis and diagnostics regulations, focusing on lateral flow assays.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service