Skip to Content
Merck
All Photos(1)

Documents

900449

Sigma-Aldrich

Graphene paper

sheet size 11.5 in. × 23.5 in., thickness 50 μm

Synonym(s):

XG Leaf® B 50 μm

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12141908

description

EMI Shielding: 53 dB @30 MHz, 58 dB @ 1.5 GHz

form

sheet

resistivity

(Electrical resistivity: surface 0.06 Ω/sq, sheet 2.7 μΩ−m)

sheet size

11.5 in. × 23.5 in.

thickness

50 μm

conductivity

(Electrical conductivity: 3700 S/cm (in-plane))
(Thermal conductivity: 550 W/mK (in-plane), 3 W/mK (through plane))

density

1.9 g/cm3

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

XG Leaf® is a more effective solution than aluminum and copper foils for a wide variety of applications to optimize thermal conductivity, thermal spreading, electrical properties, and corrosion resistance.

XG Leaf® is a thin, flexible and lightweight sheet product built on a foundation of XG
sciences, xGnP® graphene nanoplatelets. By precisely tailoring the composition, density, and
our proprietary manufacturing process, materials are created with unique properties to optimize thermal and electrical conductivity. Different types of XG Leaf® graphene paper offer outstanding thermal and electrical properties.

  • Thermal conductivity and spreading: In-plane conductivity above 550 W/mK.
  • Electrical properties: Formulations are available with surface resistivity ranging as low as 0.04 Ω/sq.

Potential applications include:
  • heat dissipation
  • resistive heating
  • EMI shielding
  • electrodes (thin battery, super capacitor, Li-air etc)
  • barrier
  • layering in composites
  • sensors

Legal Information

XG Leaf is a registered trademark of XG Sciences, Inc.
xGnP is a registered trademark of XG Sciences, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Advances in scalable synthesis and processing of two-dimensional materials

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service