Skip to Content
Merck
All Photos(2)

Key Documents

900297

Sigma-Aldrich

Polycaprolactone

viscosity 1.5 dL/g 

Synonym(s):

PCL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C6H10O2)nCH3
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

chips or granules

viscosity

1.5 dL/g

transition temp

Tm 65-70 °C (experimental)

storage temp.

−20°C

InChI

1S/C6H10O2/c7-6-4-2-1-3-5-8-6/h1-5H2

InChI key

PAPBSGBWRJIAAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professor Mitsuhiro Ebara provides insights on several types of smart nanofiber mesh systems that have been explored for different drug delivery purposes.

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service