Skip to Content
Merck
All Photos(1)

Key Documents

900079

Sigma-Aldrich

Cortisone-2,3,4-13C3 21-sulfate sodium salt solution

100 μg/mL in methanol, ≥98 atom % 13C, ≥95% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€1,220.00

€1,220.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 ML
€1,220.00

About This Item

Empirical Formula (Hill Notation):
13C3C18H27NaO8S
Molecular Weight:
465.47
UNSPSC Code:
12142200
NACRES:
NA.12

€1,220.00


Please contact Customer Service for Availability

Request more information

isotopic purity

≥98 atom % 13C

Quality Level

Assay

≥95% (CP)

form

solution

concentration

100 μg/mL in methanol

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

storage temp.

−20°C

SMILES string

O=C(COS(=O)([O-])=O)[C@@]1(O)CC[C@@]([C@]1(C)C2)([H])[C@]3([H])CCC4=[13CH][13C]([13CH2]C[C@]4(C)C3C2=O)=O.[Na+]

InChI

1S/C21H28O8S.Na/c1-19-7-5-13(22)9-12(19)3-4-14-15-6-8-21(25,17(24)11-29-30(26,27)28)20(15,2)10-16(23)18(14)19;/h9,14-15,18,25H,3-8,10-11H2,1-2H3,(H,26,27,28);/q;+1/p-1/t14-,15-,18?,19-,20-,21-;/m0./s1/i5+1,9+1,13+1;

InChI key

CKCQAVLXEMPVTM-IHWDKLTBSA-M

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service