Skip to Content
Merck
All Photos(1)

Key Documents

89377

Sigma-Aldrich

4-Fluoro-DL-glutamic acid

≥93%

Synonym(s):

(±)-2-Amino-4-fluoroglutaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8FNO4
CAS Number:
Molecular Weight:
165.12
Beilstein:
5254612
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥93%

form

solid

application(s)

peptide synthesis

SMILES string

NC(CC(F)C(O)=O)C(O)=O

InChI

1S/C5H8FNO4/c6-2(4(8)9)1-3(7)5(10)11/h2-3H,1,7H2,(H,8,9)(H,10,11)

InChI key

JPSHPWJJSVEEAX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biochemical and biological properties of methotrexate analogs containing D-glutamic acid or D-erythro, threo-4-fluoroglutamic acid.
J J McGuire et al.
Biochemical pharmacology, 42(12), 2400-2403 (1991-11-27)
Clár Donnelly et al.
Applied microbiology and biotechnology, 77(3), 699-703 (2007-09-29)
Fluorinated amino acids are used as enzyme inhibitors, mechanistic probes and in the production of pharmacologically active peptides. Because enantiomerically pure 4-fluoroglutamate is difficult to prepare, the selective degradation of the L-isomer is a potentially convenient method of obtaining D-4-fluoroglutamate
Jun Nakamura et al.
Applied and environmental microbiology, 73(14), 4491-4498 (2007-05-22)
Corynebacterium glutamicum is a biotin auxotroph that secretes L-glutamic acid in response to biotin limitation; this process is employed in industrial L-glutamic acid production. Fatty acid ester surfactants and penicillin also induce L-glutamic acid secretion, even in the presence of
D W Konas et al.
The Journal of organic chemistry, 66(26), 8831-8842 (2001-12-26)
Electrophilic fluorination of enantiomerically pure 2-pyrrolidinones (4) derived from (L)-glutamic acid has been investigated as a method for the synthesis of single stereoisomers of 4-fluorinated glutamic acids. Reaction of the lactam enolate derived from 9 with NFSi results in a
J Galivan et al.
Advances in enzyme regulation, 23, 13-23 (1985-01-01)
Methotrexate is glutamylated in cultured hepatoma cells to derivatives that contain a total of 2 to 5 gamma-glutamyl residues. The rate of polyglutamate formation and extent of accumulation are saturable with respect to both medium concentration of methotrexate and time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service