Skip to Content
Merck
All Photos(1)

Key Documents

89150

Sigma-Aldrich

Thorium nitrate hydrate

99% (T), ~4 mol/mol water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Th(NO3)4 · aq
CAS Number:
Molecular Weight:
480.06 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99% (T)

form

solid

reaction suitability

reagent type: catalyst
core: thorium

extent of labeling

3.93 kBq/g specific activity

impurities

~4 mol/mol water

SMILES string

O.[Th+4].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/4NO3.H2O.Th/c4*2-1(3)4;;/h;;;;1H2;/q4*-1;;+4

InChI key

PRFFJYANKYMDRN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Regulations require a license for the sale of radioactive substances in Switzerland. Swiss customers are kindly requested when ordering, to include their license number, issued by the Swiss Public Health Authorities.
Sales restrictions may apply.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T I Evseeva et al.
Radiatsionnaia biologiia, radioecologiia, 40(4), 456-461 (2000-10-14)
The action of 232Th-nitrate in concentration 0.09, 0.18 and 0.36 mg/l (counting on 232Th ion) on water cultures of Tradescantia (clone 02) was investigated. It was found that all investigated concentrations of 232Th showed statistically significant genotoxic effect and increased
O V Raskosha et al.
Radiatsionnaia biologiia, radioecologiia, 45(6), 744-750 (2006-02-04)
We studied the effects of both: of the separate and of the combined exposure to gamma-radiation (absorbed doses for 30 days were 1.4-1.8 cGy, for 90 days were 4.3-5.4 cGy) and 232Th nitrate (in concentrations 6.0; 20.0 and 60.0 mg/l)
Pius O Adelani et al.
Inorganic chemistry, 49(12), 5701-5705 (2010-05-15)
The hydrothermal reactions of thorium nitrate and uranyl acetate with carboxyphenylphosphonic acid and HF result in the formation of ThF(2)(PO(3)C(6)H(4)CO(2)H) and UO(2)(PO(3)HC(6)H(4)CO(2)H)(2).2H(2)O, respectively. ThF(2)(PO(3)C(6)H(4)CO(2)H) adopts a pillared structure constructed from thorium oxyfluoride layers built from [ThO(4)F(4)] units that are bridged
G N Stradling et al.
Human & experimental toxicology, 14(2), 165-169 (1995-02-01)
1. With DTPA as a comparison, the siderophore analogue 3,4,3-LIHOPO has been examined for its ability to remove 228Th nitrate from the rat after subcutaneous (sc) and intramuscular (im) injection to simulate wound contamination. The commencement of treatment was delayed
V Jobbágy et al.
Radiation protection dosimetry, 141(4), 440-443 (2010-09-11)
Thoron and progeny are decay products of (232)Th with a great impact on human health. The release of thoron gas from the mining and milling of thorite, monazite and other major thorium ores has been recognised as a potential radiological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service