Skip to Content
Merck
All Photos(1)

Key Documents

88482

Sigma-Aldrich

1-Butyl-4-methylpyridinium chloride

≥97.0%

Synonym(s):

1-Butyl-4-picolinium chloride, 4MBPCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16ClN
CAS Number:
Molecular Weight:
185.69
Beilstein:
3734338
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Pricing and availability is not currently available.

Assay

≥97.0%

mp

158-160 °C

SMILES string

[Cl-].CCCC[n+]1ccc(C)cc1

InChI

1S/C10H16N.ClH/c1-3-4-7-11-8-5-10(2)6-9-11;/h5-6,8-9H,3-4,7H2,1-2H3;1H/q+1;/p-1

InChI key

YTSDTJNDMGOTFN-UHFFFAOYSA-M

Other Notes

Linear dimerization of 1-alkenes in a 4-MBP Cl- - AlCl3system[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B. Ellis et al.
Chemical Communications (Cambridge, England), 337-337 (1999)
Viresh R Thamke et al.
Environmental pollution (Barking, Essex : 1987), 250, 567-577 (2019-04-27)
The present study deals with the cyto-genotoxicological impact of ionic liquids, 1-butyl-3-methylimidazolium bromide, trihexyl tetradecylphosphonium dicyanamide, 1-decyl-3-methylimidazolium tetrafluoroborate, benzyldimethyltetradecylammonium chloride, and 1-butyl-4-methylpyridinium chloride, on animal cells and their biodegradation. The long alkyl chain containing ionic liquids were found to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service