Skip to Content
Merck
All Photos(3)

Key Documents

86864

Sigma-Aldrich

Tetrabutylammonium cyanate

technical

Synonym(s):

Tetrabutylammonium isocyanate, Tetrabutylammonium oxycyanide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€294.00
25 G
€473.00

€294.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€294.00
25 G
€473.00

About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(OCN)
CAS Number:
Molecular Weight:
284.48
Beilstein:
5671059
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€294.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

technical

form

crystals

storage temp.

2-8°C

SMILES string

[O-]C#N.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.CHNO/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;2-1-3/h5-16H2,1-4H3;3H/q+1;/p-1

InChI key

PEFQHPVDKJBRFF-UHFFFAOYSA-M

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service