Recommended Products
Assay
98%
form
solid
optical activity
[α]25/D +40.2°, c = 2 in ethanol
reaction suitability
reaction type: solution phase peptide synthesis
mp
76-78 °C (lit.)
application(s)
peptide synthesis
storage temp.
2-8°C
SMILES string
OC(=O)[C@H]1CCCN1C(=O)OCc2ccccc2
InChI
1S/C13H15NO4/c15-12(16)11-7-4-8-14(11)13(17)18-9-10-5-2-1-3-6-10/h1-3,5-6,11H,4,7-9H2,(H,15,16)/t11-/m1/s1
InChI key
JXGVXCZADZNAMJ-LLVKDONJSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service