Skip to Content
Merck
All Photos(1)

Key Documents

860395

Sigma-Aldrich

(1R,2S)-(−)-Ephedrine nitrate

99%

Synonym(s):

(1R,2S)-(−)-α-(1-Methylaminoethyl)benzyl alcohol nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH[CH(NHCH3)CH3]OH·HNO3
CAS Number:
Molecular Weight:
228.25
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

99%

optical activity

[α]21/D −31°, c = 5 in H2O

mp

126-128 °C (lit.)

SMILES string

O[N+]([O-])=O.CN[C@@H](C)[C@H](O)c1ccccc1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service