Skip to Content
Merck
All Photos(1)

Key Documents

854980

Sigma-Aldrich

2′-Deoxycytidine hydrate

≥99% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O4 · xH2O
CAS Number:
Molecular Weight:
227.22 (anhydrous basis)
Beilstein:
87567
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99% (HPLC)

form

crystals

optical activity

[α]25/D +54°, c = 2 in H2O

mp

209-211 °C (lit.)

SMILES string

O.NC1=NC(=O)N(C=C1)[C@H]2C[C@H](O)[C@@H](CO)O2

InChI

1S/C9H13N3O4.H2O/c10-7-1-2-12(9(15)11-7)8-3-5(14)6(4-13)16-8;/h1-2,5-6,8,13-14H,3-4H2,(H2,10,11,15);1H2/t5-,6+,8+;/m0./s1

InChI key

HXBGOHZLZCFWLH-OERIEOFYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Joshua J Klaene et al.
Journal of chromatography. A, 1439, 112-123 (2015-11-27)
LC-MS using electrospray ionization is currently the method of choice in bio-organic analysis covering a wide range of applications in a broad spectrum of biological media. The technique is noted for its high sensitivity but one major limitation that hinders

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service