Skip to Content
Merck
All Photos(1)

Key Documents

85271

Sigma-Aldrich

Silver perchlorate hydrate

≥96.0% (calc. based on dry substance, AT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgClO4 · xH2O
CAS Number:
Molecular Weight:
207.32 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder or crystals

grade

for analytical purposes

Assay

≥96.0% (calc. based on dry substance, AT)

form

powder or crystals

reaction suitability

reagent type: oxidant

impurities

≤12% water

SMILES string

O.[Ag+].[O-]Cl(=O)(=O)=O

InChI

1S/Ag.ClHO4.H2O/c;2-1(3,4)5;/h;(H,2,3,4,5);1H2/q+1;;/p-1

InChI key

IXFNJWPLHOWEPT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service