Skip to Content
Merck
All Photos(1)

Documents

852686

Sigma-Aldrich

6-Mercaptopurine-9-β-D-ribofuranoside

≥95.0% (HPLC)

Synonym(s):

6-Mercaptopurine riboside, 6-Purinethiol riboside, 6-Thioinosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N4O4S
CAS Number:
Molecular Weight:
284.29
Beilstein:
40536
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95.0% (HPLC)

form

powder

optical activity

[α]/D −74±4°, c = 1% in 0.1 M NaOH

mp

220-223 °C (lit.)

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(S)ncnc23

InChI

1S/C10H12N4O4S/c15-1-4-6(16)7(17)10(18-4)14-3-13-5-8(14)11-2-12-9(5)19/h2-4,6-7,10,15-17H,1H2,(H,11,12,19)/t4-,6-,7-,10-/m1/s1

InChI key

NKGPJODWTZCHGF-KQYNXXCUSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lennard P L Gilissen et al.
Journal of Crohn's & colitis, 6(6), 698-707 (2012-03-09)
Therapeutic drug monitoring of active metabolites of thiopurines, azathioprine and 6-mercaptopurine, is relatively new. The proposed therapeutic threshold level of the active 6-thioguanine nucleotides (6-TGN) is ≥235 pmol/8×10(8) erythrocytes. The aim of this prospective cross-sectional study was to compare 6-TGN
Sumit Parmar et al.
Pharmacogenomics, 12(4), 503-514 (2011-04-28)
To adopt an individualized approach to assess cytarabine (ara-C) hematotoxicity, we studied the relationship between pharmacogenetic variability in the cytidine deaminase gene (CDA) and ara-C toxicity in native peripheral blood mononuclear cells from 100 healthy volunteers. Peripheral blood mononuclear cells
Anwar Saad Abd-Elfattah et al.
The Journal of thoracic and cardiovascular surgery, 144(1), 250-255 (2012-02-15)
To determine the role of the p-nitrobenzylthioinosine-sensitive equilibrative nucleoside transporter 1 (es-ENT1) in postmyocardial infarction reperfusion injury-mediated ventricular fibrillation and regional dysfunction. We used erythro-9 (2-hydroxy-3-nonyl)-adenine and p-nitrobenzylthioinosine to inhibit both adenosine deamination and transport in a canine model of
Daniel Pecher et al.
Molecules (Basel, Switzerland), 23(7) (2018-07-19)
Thiopurines (TP) represent an important therapeutic tool for the treatment of inflammatory bowel diseases (IBD) in the current situation of rising incidence and health care costs. The results of multiple clinical studies aimed at finding correlations between levels of TP
Microwave-promoted "one-pot" synthesis of 4-nitrobenzylthioinosine analogues using thiourea as a sulfur precursor.
Hong-Ying Niu et al.
Chemistry, an Asian journal, 7(1), 45-49 (2011-10-18)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service