Skip to Content
Merck
All Photos(1)

Documents

82340

Sigma-Aldrich

Propylhydrazine oxalate salt

≥98.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2NHNH2 · (COOH)2
CAS Number:
Molecular Weight:
164.16
Beilstein:
3703104
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

form

crystals

SMILES string

CCCNN.OC(=O)C(O)=O

InChI

1S/C3H10N2.C2H2O4/c1-2-3-5-4;3-1(4)2(5)6/h5H,2-4H2,1H3;(H,3,4)(H,5,6)

InChI key

ATXLENZWWDUZLH-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetyana Kobets et al.
Mutation research, 844, 10-24 (2019-07-22)
DNA-damaging activities of twenty-four structurally diverse unsubstituted and substituted cyclic compounds were assessed in embryo-fetal chicken livers. Formation of DNA adducts and strand breaks were measured using the nucleotide 32P-postlabelling (NPL) and comet assays, respectively. Unsubstituted monocyclic benzene, polycyclic fused

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service