Skip to Content
Merck
All Photos(1)

Key Documents

809144

Sigma-Aldrich

Polyphthalaldehyde

Cyclic

Synonym(s):

cPPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C8H6O2)n
UNSPSC Code:
12352200

form

powder

mol wt

average Mn 10,000-20,000

PDI

<3

Looking for similar products? Visit Product Comparison Guide

Application

PPA is a self-amplified depolymerization media for dry lithography.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hector Lopez Hernandez et al.
Advanced materials (Deerfield Beach, Fla.), 26(45), 7637-7642 (2014-10-22)
Triggerable transient electronics are demonstrated with the use of a metastable poly(phthalaldehyde) polymer substrate and encapsulant. The rate of degradation is controlled by the concentration of the photo-acid generator and UV irradiance. This work expands on the materials that can
Joshua A Kaitz et al.
Journal of the American Chemical Society, 135(34), 12755-12761 (2013-08-09)
End-capped poly(phthalaldehyde) (PPA) synthesized by anionic polymerization has garnered significant interest due to its ease of synthesis and rapid depolymerization. However, alternative ionic polymerizations to produce PPA have been largely unexplored. In this report, we demonstrate that a cationic polymerization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service