Skip to Content
Merck
All Photos(6)

Documents

805424

Sigma-Aldrich

Reduced graphene oxide

Synonym(s):

rGO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CxOyHz
UNSPSC Code:
12352103
NACRES:
NA.23

description

chemically reduced by hydrizine

form

powder

composition

Carbon, >75%
Nitrogen, <5%

surface area

103 m2/g

conductivity

7111 S/m (pressed pallet)

Looking for similar products? Visit Product Comparison Guide

General description

Chemically reduced by hydrazine.

Application

Hydrazine-reduced graphene oxide can be used as microwave absorbing material. Other applications include:
  • Supercapacitors
  • Dye-sensitized solar cells and organic solar cells
  • Batteries
  • Drug delivery

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

805424
Wang C, et al.
Applied Physics Letters, 98(7), 072906-072906 (2011)
Nickel-Cobalt Layered Double Hydroxide Nanosheets for High-performance Supercapacitor Electrode Materials
Chen, et al.
Advances in Functional Materials, 24(7), 934-942 (2014)
Chun Kiang Chua et al.
Chemical Society reviews, 43(1), 291-312 (2013-10-15)
The chemical reduction of graphene oxide is a promising route towards the large scale production of graphene for commercial applications. The current state-of-the-art in graphene oxide reduction, consisting of more than 50 types of reducing agent, will be reviewed from
Daniel R Dreyer et al.
Chemical Society reviews, 39(1), 228-240 (2009-12-22)
The chemistry of graphene oxide is discussed in this critical review. Particular emphasis is directed toward the synthesis of graphene oxide, as well as its structure. Graphene oxide as a substrate for a variety of chemical transformations, including its reduction

Articles

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Advances in scalable synthesis and processing of two-dimensional materials

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service