Skip to Content
Merck
All Photos(1)

Key Documents

805041

Sigma-Aldrich

2-(trimethylsilyl)-2-cyclohexen-1-one

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€142.00

€142.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€142.00

About This Item

Empirical Formula (Hill Notation):
C9H16OSi
CAS Number:
Molecular Weight:
168.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€142.00


Please contact Customer Service for Availability

Request a Bulk Order

description

Fp: 15.8°F

Quality Level

form

liquid

refractive index

n20/D 1.4524

density

0.8680 g/mL at 25 °C

functional group

ketone

storage temp.

−20°C

SMILES string

O=C1C([Si](C)(C)C)=CCCC1

InChI

1S/C9H16OSi/c1-11(2,3)9-7-5-4-6-8(9)10/h7H,4-6H2,1-3H3

InChI key

SXGCQSJRVVKTOH-UHFFFAOYSA-N

Related Categories

General description

2-(Trimethylsilyl)-2-cyclohexen-1-one is a silyl enone.

Application

The following silyl enone can be converted into a highly useful cyclohexyne precursor as reported by the Garg and DuBois groups. The resulting silyltriflate adduct can be subjected to various dienes and dipolarophiles in the presence of Cesium Fluoride.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

15.8 °F

Flash Point(C)

-9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service