Skip to Content
Merck
All Photos(1)

Documents

803901

Sigma-Aldrich

PtCo/reduced graphene oxide nanocomposite

10 mg/mL, dispersion in acetone

Synonym(s):

PtCo Alloy Decorated Reduced Graphene Oxide, PtCo/RGO Nanocomposite

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

description

Loading Density: 10%- 50% (PtCo nanocrystal) by TEM
Resistance: <103 Ω/sq (graphene)

Quality Level

form

dispersion

composition

acetone, ~80%
PtCo nanocrystal, 2-10%
reduced graphene oxide, 10-18%

concentration

10 mg/mL

thickness

1-10 nm , graphene

particle size

2-5 nm (PtCo nanocrystal)

storage temp.

2-8°C

Application

Efficient electrocatalyst material for fuel cells including methanol oxidation and proton exchange membrane.

Legal Information

Product of Ocean Nanotech, LLC.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F

Flash Point(C)

5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of Graphene Supported Platinum-Cobalt Nanocomposites as Electrocatalysts for Ethanol Oxidation.
Kepeniene V, et al.
Journal of the Electrochemical Society, 161(14), F1354-F1359 (2014)
Monodispersed PtCo nanoparticles on hexadecyltrimethylammonium bromide treated graphene as an effective oxygen reduction reaction catalyst for proton exchange membrane fuel cells.
Nam et al.
Carbon, 50(10), 3739-3747 (2012)

Articles

Graphene is a unique two-dimensional (2D) structure of monolayer carbon atoms packed into a dense honeycomb crystal that has attracted great interest due to its diverse and fascinating properties.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service