Skip to Content
Merck
All Photos(1)

Documents

803146

Sigma-Aldrich

Hydrocortisone-2,3,4-13C3 solution

100 μg/mL in methanol, 99 atom % 13C, 98% (CP)

Synonym(s):

Cortisol-2,3,4-13C3 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C18H30O5
Molecular Weight:
365.44
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

Assay

98% (CP)

form

liquid

concentration

100 μg/mL in methanol

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@@]12C(CC[C@]2(O)C(CO)=O)C3CCC4=[13CH][13C]([13CH2]C[C@]4(C)C3[C@@H](O)C1)=O

InChI

1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14?,15?,16-,18?,19-,20-,21-/m0/s1/i5+1,9+1,13+1

InChI key

JYGXADMDTFJGBT-KKPOWBTRSA-N

Related Categories

Application

This product may be used as an analytical standard.

Packaging

1 mL in ampule
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyun Wu et al.
Cytotherapy, 22(7), 377-387 (2020-05-23)
The use of animal serum in culture medium brings safety concerns and batch-to-batch variability, and thus may restrict the clinical use of ex vivo expanded mesenchymal stromal cells (MSCs). Clinically compliant MSCs should be developed in adherence to serum-free, xeno-free

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service