Skip to Content
Merck
All Photos(1)

Key Documents

799661

Sigma-Aldrich

Fe/ppm Pd for Suzuki-Miyaura Cross-Couplings

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12161600

form

powder, crystals or chunks

reaction suitability

core: iron
reaction type: Cross Couplings
reagent type: catalyst

Application

Nanoparticles (NPs) of this material contain parts per million (ppm) levels of Pd. The NPs can catalyze Suzuki-Miyaura (SM) reactions, including cases involving highly challenging reaction partners. Nano micelles are employed to both solubilize and deliver the reaction partners to the Fe/ppm Pd catalyst resulting in C-C bond formation.

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sachin Handa et al.
Science (New York, N.Y.), 349(6252), 1087-1091 (2015-09-05)
Most of today's use of transition metal-catalyzed cross-coupling chemistry relies on expensive quantities of palladium (Pd). Here we report that nanoparticles formed from inexpensive FeCl3 that naturally contains parts-per-million (ppm) levels of Pd can catalyze Suzuki-Miyaura reactions, including cases that
When Does Organic Chemistry Follow Nature?s Lead and ?Make the Switch?.
Lipshutz B H
The Journal of Organic Chemistry, 82(6), 2806-2816 (2017)
New surfactants for chemistry in water.
Krause N
Current opinion in green and sustainable chemistry., 7, 18-22 (2017)
Switching from organic solvents to water at an industrial scale.
Parmentier M, et al.
Current opinion in green and sustainable chemistry., 7, 13-17 (2017)

Related Content

Designer surfactants enable diverse transformations like Suzuki-Miyaura, Olefin Metathesis, and 1,4-Addition to Enones in water by Prof. Bruce Lipshutz.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service