Skip to Content
Merck
All Photos(1)

Documents

798959

Sigma-Aldrich

cis-2-Bromovinylboronic acid MIDA ester

90%

Synonym(s):

MIDA006

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9BBrNO4
CAS Number:
Molecular Weight:
261.87
UNSPSC Code:
12352103
NACRES:
NA.22

Assay

90%

form

solid

mp

153-159 °C (Decomp.)

storage temp.

−20°C

SMILES string

CN1CC(=O)OB(OC(=O)C1)\C=C/Br

InChI

1S/C7H9BBrNO4/c1-10-4-6(11)13-8(2-3-9)14-7(12)5-10/h2-3H,4-5H2,1H3/b3-2-

InChI key

ALXOFLXCBIRRRC-IHWYPQMZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The (Z)-(2-Bromovinyl)-MIDA boronate is a unique building block for the incorporation of cis-olefin for small molecule synthesis. Burke and coworkers have displayed the extreme versatility of this building block in the construction of polyene-containing natural product motifs.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service