Skip to Content
Merck
All Photos(1)

Key Documents

798126

Sigma-Aldrich

Poly(D,L-lactide-block-acrylic acid)

Mn PDLLA:PAA 5,000:18,000, PDI <1.3

Synonym(s):

PDLLA -b-PAA 5kDa, PLA-PAA, Polylactide-block-polyacrylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C13H25S3(C3H4O2)xC6H9NO(C3H4O2)yH
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

mol wt

PAA Mn 18,000
PDLLA Mn 5,000

PDI

<1.3

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

This diblock copolymer can be used to self-assemble into a drug delivery vehicle. This particular block ratio and molecular weight forms spherical micelles.

Legal Information

Sold for research purposes only: See sigma-aldrich.com/raftlicense Patents: WO98/01478, WO99/311444.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service