Skip to Content
Merck
All Photos(1)

Key Documents

797375

Sigma-Aldrich

4,4′,4′′,4′′′-(Ethene-1,1,2,2-tetrayl)tetraphenol

Synonym(s):

4,4′,4′′,4′′′-(1,2-Ethenediylidene)tetrakisphenol, TPE-TOH, Tetra(4-hydroxyphenyl)ethylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H20O4
CAS Number:
Molecular Weight:
396.43
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

SMILES string

OC(C=C1)=CC=C1C(C2=CC=C(O)C=C2)=C(C3=CC=C(O)C=C3)C4=CC=C(O)C=C4

InChI

1S/C26H20O4/c27-21-9-1-17(2-10-21)25(18-3-11-22(28)12-4-18)26(19-5-13-23(29)14-6-19)20-7-15-24(30)16-8-20/h1-16,27-30H

InChI key

QQUZHNPGWNIYMK-UHFFFAOYSA-N

Application

TPE-TOH is a synthetic intermediate of aggregation-induced emission (AIE) dye for use in further synthesis of alkyl-halogen to make ether via esterification and polymer reaction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile synthesis of soluble nonlinear polymers with glycogen-like structures and functional properties from ?simple? acrylic monomers
Rongrong Hu; Jacky W. Y. Lam; Yong Yu; Herman H. Y. Sung; Ian D. Williams; et al
Polym. Chem., 4, 95-95 (2013)

Articles

AIE Luminogens - Family of new materials with multifaceted functionalities. Find some representative applications of the AIEgens

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service