Skip to Content
Merck
All Photos(1)

Documents

795712

Sigma-Aldrich

Molybdenum tris(1,2-bis(trifluoromethyl)ethane-1,2-dithiolene)

Synonym(s):

Mo(tfd)3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12F18MoS6
CAS Number:
Molecular Weight:
774.43
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

SMILES string

FC(F)(F)C(S1)=C(C(F)(F)F)S[Mo]213(SC(C(F)(F)F)=C(C(F)(F)F)S3)SC(C(F)(F)F)=C(C(F)(F)F)S2

InChI

1S/3C4H2F6S2.Mo/c3*5-3(6,7)1(11)2(12)4(8,9)10;/h3*11-12H;/q;;;+6/p-6/b3*2-1-;

InChI key

KPELOYUGUICPOT-JVUUZWNBSA-H

General description

The p dopant Mo(tfd)3 has high electron affinity (EA= 5.6 eV).The efficiency of Mo(tfd)3 as a dopant has been studied extensively by ultraviolet and X ray photoelectron spectroscopy, Rutherford backscattering, etc.

Application

Molybdenum tris(1,2-bis(trifluoromethyl)ethane-1,2-dithiolene) (Mo(tfd)3) may be used to p dope the hole transporting layer of HTM N,N′-di[(1-naphthyl)-N,N′-diphenyl]-1,1′-biphenyl-4,4′- diamine.
It may be used as a dopant to reduced contact resistance in pentacene based organic field effect transistors (OFETs).

Biochem/physiol Actions

p-dopant

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pentacene organic field-effect transistors with doped electrode-semiconductor contacts
Tiwari SP, et al.
Organic Electronics null
A Molybdenum Dithiolene Complex as p-Dopant for Hole-Transport Materials: A Multitechnique Experimental and Theoretical Investigation
Qi, Y.;
Chemistry of Materials, 22, 524-524 (2010)
Yabing Qi et al.
Journal of the American Chemical Society, 131(35), 12530-12531 (2009-08-15)
Experimental and theoretical results are presented on the electronic structure of molybdenum tris[1,2-bis(trifluoromethyl) ethane-1,2-dithiolene] (Mo(tfd)(3)), a high electron-affinity organometallic complex that constitutes a promising candidate as a p-dopant for organic molecular semiconductors. The electron affinity of the compound, determined via
Use of a high electron-affinity molybdenum dithiolene complex to p-dope hole-transport layers.
Qi Y, et al.
Journal of the American Chemical Society, 12530-12531 null
Remote doping of a pentacene transistor: Control of charge transfer by molecular-level engineering
Zhao, W.;
Applied Physics Letters, 97, 123305/1-123305/1 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service