Skip to Content
Merck
All Photos(1)

Key Documents

794465

Sigma-Aldrich

Ritter Pentafluoroiodoethane-TMG Reagent

Synonym(s):

TMG-CF3CF2I, Tetramethylguanidine-pentafluoroethyl iodide adduct

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13F5IN3
Molecular Weight:
361.09
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available. Contact Technical Service

form

liquid

reaction suitability

reaction type: C-C Bond Formation

density

1.53 g/mL

functional group

amine
fluoro
iodo

shipped in

dry ice

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)I.N=C(N(C)C)N(C)C

InChI

1S/C5H13N3.C2F5I/c1-7(2)5(6)8(3)4;3-1(4,5)2(6,7)8/h6H,1-4H3;

InChI key

ZFIASFCSDAHOLE-UHFFFAOYSA-N

Application

Product is a more convenient, liquified version of pentafluoroiodoethane, which can be utilized in photocatalytic and electrophilic pentafluoroethylation of olefins and heteroatoms respectively.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Ritter lab currently focuses on fluorination chemistry for late-stage functionalization of complex natural and unnatural products. PhenoFluor™ has been developed as a general reagent for the selective, predictable, direct deoxyfluorination of complex alcohols and phenols.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service