Skip to Content
Merck
All Photos(1)

Key Documents

793833

Sigma-Aldrich

Methylammonium triiodoplumbate(II) precursor solution

greener alternative

40 wt. % in DMF

Synonym(s):

Methanamine, triiodoplumbate(1−), Methylamine triiodoplumbate(1−), Methylammonium triiodoplumbate(1−), Plumbate(1−), triiodo-, hydrogen, compound with methanamine (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3NH3]+[PbI3]-
Molecular Weight:
619.98
UNSPSC Code:
12352302

form

solution

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

40 wt. % in DMF

refractive index

n20/D 1.520

density

1.368 g/mL at 25 °C

λmax

321 nm

greener alternative category

Application

Methylammonium lead iodide is a well studied precursor for the deposition of perovskite active layers for advanced photovoltaics

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

141.0 °F

Flash Point(C)

60.56 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nam Joong Jeon et al.
Journal of the American Chemical Society, 135(51), 19087-19090 (2013-12-10)
A set of three N,N-di-p-methoxyphenylamine-substituted pyrene derivatives have successfully been synthesized and characterized by (1)H/(13)C NMR spectroscopy, mass spectrometry, and elemental analysis. The optical and electronic structures of the pyrene derivatives were adjusted by controlling the ratio of N,N-di-p-methoxyphenylamine to

Articles

Solar panel demand surges for sustainable energy, with power output rising from 5.0 to 90 GWh/d from 2010 to 2016.

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Controlled synthesis of metal clusters regulates ligands and atoms, advancing metal nanomaterial synthesis.

Next generation solar cells have the potential to achieve conversion efficiencies beyond the Shockley-Queisser (S-Q) limit while also significantly lowering production costs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service