Skip to Content
Merck
All Photos(1)

Key Documents

792020

Sigma-Aldrich

Poly(D,L-lactide), 4-cyano-4-[(dodecylsulfanylthiocarbonyl)sulfanyl]pentonate terminated

average Mn 5000, PDI ≤1.5

Synonym(s):

PDLLA RAFT terminated, RAFT-PLA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H35NOS3(C3H4O2)n
UNSPSC Code:
12162002

form

solid

mol wt

average Mn 5000

PDI

≤1.5

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Polylactide is commonly used in drug delivery applications. The RAFT functionality can be used to generate a second block to create an amphiphilic diblock copolymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service