Skip to Content
Merck
All Photos(1)

Key Documents

78055

Sigma-Aldrich

L-Phenylalanine allyl ester p-toluenesulfonate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO2 · C7H8O3S
CAS Number:
Molecular Weight:
377.45
Beilstein:
5212778
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

optical activity

[α]20/D +10.5±1°, c = 1% in DMF

mp

155-158 °C

application(s)

peptide synthesis

SMILES string

Cc1ccc(cc1)S(O)(=O)=O.N[C@@H](Cc2ccccc2)C(=O)OCC=C

InChI

1S/C12H15NO2.C7H8O3S/c1-2-8-15-12(14)11(13)9-10-6-4-3-5-7-10;1-6-2-4-7(5-3-6)11(8,9)10/h2-7,11H,1,8-9,13H2;2-5H,1H3,(H,8,9,10)/t11-;/m0./s1

InChI key

PKXPRZFLCJFMHN-MERQFXBCSA-N

Other Notes

Used in peptide and N-glycopeptide synthesis; the ester is selectively deblocked with (Ph3P)3RhCl[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Waldmann et al.
Liebigs Ann. Chem., 1712-1712 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service