Skip to Content
Merck
All Photos(1)

Key Documents

77939

Sigma-Aldrich

(R)-3,3′-Di-tert-butyl-5,5′,6,6′,7,7′,8,8′-octahydro-1,1′-(bi-2-naphthol) dipotassium salt

purum, ≥95.0% (dry substance, CHN)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H36K2O2
CAS Number:
Molecular Weight:
482.78
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

purum

Assay

≥95.0% (dry substance, CHN)

form

powder

impurities

<10% water

SMILES string

[K+].[K+].CC(C)(C)c1cc2CCCCc2c(c1[O-])-c3c([O-])c(cc4CCCCc34)C(C)(C)C

InChI

1S/C28H38O2.2K/c1-27(2,3)21-15-17-11-7-9-13-19(17)23(25(21)29)24-20-14-10-8-12-18(20)16-22(26(24)30)28(4,5)6;;/h15-16,29-30H,7-14H2,1-6H3;;/q;2*+1/p-2

InChI key

KUFMCCDFHGQYSN-UHFFFAOYSA-L

Application

Octahydro-BINOL was demonstrated to be an effective ligand in Mo-catalyzed desymmetrization of a triene by asymmetric ring closing metathesis.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

A readily available user-friendly chiral catalyst for efficient enantioselective olefin metathesis, used in conjunction with Cat. No. 75287, a molybdenum complex

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah J Dolman et al.
Journal of the American Chemical Society, 124(24), 6991-6997 (2002-06-13)
The first catalytic asymmetric ring-closing metathesis method for the synthesis of N-containing heterocycles is reported; this is accomplished through Mo-catalyzed kinetic resolution or desymmetrization of unsaturated amines. Importantly, this catalytic asymmetric method delivers medium-ring unsaturated amines (including eight-membered rings) in
A Readily Available and User-Friendly Chiral Catalyst for Efficient Enantioselective Olefin Metathesis This research was supported by the National Institutes of Health (GM-59426) and the National Science Foundation (CHE-9905806 to A. H. H. and CHE-9988766 to R. R. S.).
Sarah L. Aeilts et al.
Angewandte Chemie (International ed. in English), 40(8), 1452-1456 (2001-04-24)
S.L. Aeilts et al.
Angewandte Chemie (International Edition in English), 113, 1500-1500 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service