Skip to Content
Merck
All Photos(1)

Key Documents

779385

Sigma-Aldrich

3,4-Dihydroisoquinoline

≥97.5% (GC)

Synonym(s):

3,4-Dihydroisoquinoline

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€166.00
5 G
€828.00

€166.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€166.00
5 G
€828.00

About This Item

Empirical Formula (Hill Notation):
C9H9N
CAS Number:
Molecular Weight:
131.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€166.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥97.5% (GC)
97.5-102.5% (T)

form

solid

suitability

complies for identity (IR)

SMILES string

C1Cc2ccccc2C=N1

InChI

1S/C9H9N/c1-2-4-9-7-10-6-5-8(9)3-1/h1-4,7H,5-6H2

InChI key

NKSZCPBUWGZONP-UHFFFAOYSA-N

Application

3,4-Dihydroisoquinoline can be used as a reactant to synthesize:
  • 5,6-Dihydro-8H-isoquino[1,2-b]quinazolin-8-one by decarboxylative cyclization reaction with isatoic anhydride using tetrabutylammonium iodide (TBAI).[1]
  • 1-naphtholyl tetrahydroisoquinoline by aza-Friedel-Crafts reaction with various naphthols.[2]
  • 3,4-dihydroisoquinoline pseudo bases, which are employed as starting materials for the preperation of 3-benzazepine derivatives.[3]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrosynthesis of polycyclic quinazolinones and rutaecarpine from isatoic anhydrides and cyclic amines
Chen Xingyu, et al.
Royal Society of Chemistry Advances, 10(72), 44382-44386 (2020)
A facile, one pot method for the synthesis of 4-acyl-1, 2-dihydro-3-benzazepines, based on the ring expansion of natural and synthetic 3, 4-dihydroisoquinoline pseudo bases
Kartsev VG, et al.
Tetrahedron Letters, 56(50), 6988-6993 (2015)
Solvent-free direct aza-Friedel-Crafts reactions between 3, 4-dihydroisoquinoline and 1-or 2-naphthols
MacLeod PD, et al.
Tetrahedron Letters, 47(38), 6791-6794 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service