Skip to Content
Merck
All Photos(1)

Key Documents

778532

Sigma-Aldrich

(SP)-1-[(R)-1-(Di-tert-butylphosphino)ethyl]-2-[(R)-phenylphosphinoyl]ferrocene

97%

Synonym(s):

(2R)-1-[(1R)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-[(R)-phenylphosphinyl]ferrocene (acc to CAS), JoSPOphos SL-J688-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H36FeOP2
CAS Number:
Molecular Weight:
482.36
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥97%
97%

form

powder

optical purity

ee: ≥99%

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@H]([C]2[CH][CH][CH][C]2[P@@H](=O)c3ccccc3)P(C(C)(C)C)C(C)(C)C

InChI

1S/C21H31OP2.C5H5.Fe/c1-16(24(20(2,3)4)21(5,6)7)18-14-11-15-19(18)23(22)17-12-9-8-10-13-17;1-2-4-5-3-1;/h8-16,23H,1-7H3;1-5H;/t16-;;/m1../s1

InChI key

ZRFUSACJLNCGTO-GGMCWBHBSA-N

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral mixed secondary phosphine-oxide-phosphines: high-performing and easily accessible ligands for asymmetric hydrogenation.
Heidi Landert et al.
Angewandte Chemie (International ed. in English), 49(38), 6873-6876 (2010-08-18)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service